Skip to Content
Merck
All Photos(2)

Key Documents

1.46703

Millipore

Sabouraud Dextrose Agar

Lecithin, Tween®, lid, ICR plus: lockable settle plates, diam. 90 mm, sterile; γ-irradiated, triple packed of 20 and 120 plates, suitable for surface monitoring

Synonym(s):

ICR plus settle plate, SDA, SDA Settle Plate, SDA with neutralizers

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$117.00
120 EACH
$551.00

$117.00


Estimated to ship on20 May 2025


Request more information

Select a Size

Change View
20 EA
$117.00
120 EACH
$551.00

About This Item

UNSPSC Code:
41106227
NACRES:
NA.74

$117.00


Estimated to ship on20 May 2025


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

lid ICR plus: lockable settle plates
ready-to-use

composition

Lecithin
Tween®

packaging

triple packed of 20 and 120 plates

diam.

90 mm

color

clear yellowish Medium

pH range

5.4-5.8

application(s)

air monitoring
pharmaceutical

surface monitoring

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Agar + LT – ICR+ settle plate is a selective media for the determination of the total count of yeasts and molds. Settle plates are used for active or passive air monitoring as well for fingerprints of personnel. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favours fungal growth and Lecithin and polysorbate (Tween®) 80 are neutralizers to inactivate residues of disinfectants. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport of the samples without the risk of losing the lid. For anaerobic incubation the lid has to be turned into the vent position.

Application

Sabouraud Dextrose Agar (SDA) is a selective medium for cultivation and isolation of yeasts and molds. The media is used for environmental monitoring by determining the yeast and mold count in air by active or passive air monitoring as well as fingerprints of personnel.

Storage and Stability

  • The product can be used for sampling until the expiry date if stored upright, protected from light, and properly sealed at +15 °C to +25 °C.
  • Condensation can be prevented by avoiding quick temperature shifts and mechanical stress.
  • The testing procedures described on the CoA can be started up to the expiry date printed on the label.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service