Skip to Content
Merck
All Photos(1)

Key Documents

1.46496

Millipore

Modified Tween®-Letheen Broth

ready-to-use, bottle volume 90 mL , Filling volume

Synonym(s):

MLBT, Modified Letheen Broth with Tween®, MTLB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214
NACRES:
NA.74
Pricing and availability is not currently available.

description

For enumeration of fastidious microorganisms in the presence of disinfectant residues

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

6 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

bottle of
pkg of (90 ml in 125 ml bottle with blue screw cap and 3 loci (10 bottles per box))

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

125 mL

bottle volume

90 mL , Filling volume

pH

7.2(ready-to-use solution)

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

components

histidine
tween
lecithin
sodium thiosulfate

storage temp.

2-25°C

suitability

nonselective for

General description

Modified Tween®-Letheen Broth has a significantly improved nutritional content when compared to the Letheen-Broth. This has higher meat peptone concentration along with casein and yeast extract vitamin sources. Addition of sodium thiosulfate (neutralizes iodine and chlorine) and sodium bisulfite (neutralizes formaldehyde, formaldehyde releasers and Glutaraldehyde) has improved the efficacy of the inactivating agents.

Application

Modified Tween®-Letheen-Broth is designed for the enumeration of fastidious microorganisms in the presence of disinfectant residues.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service