Skip to Content
Merck
All Photos(1)

Key Documents

1.46492

Millipore

Tryptic Soy Broth Ready-to-use Bottles

cysteine (Inactivator B.), Histidine, Tween®, Lecithin, bottle capacity 125 mL, bottle filling volume 90 mL, closure type, blue screw cap with 3 loci, box of 10 bottles

Synonym(s):

CASO, Casein Soybean Digest Broth with 3% Tween, 0.3% Lecithin, 0.1% Histidine and 0.1% Cysteine, SCDB, Tryptic Soy Broth with TLHC (Inactivator B.), Tryptic Soy Broth with neutralizer

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 90 ML
$101.00

$101.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
10 X 90 ML
$101.00

About This Item

UNSPSC Code:
41106226
NACRES:
NA.74

$101.00


Estimated to ship on29 May 2025


Request more information

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type blue screw cap with 3 loci
ready-to-use

composition

cysteine (Inactivator B.)
Histidine
Lecithin
Tween®

packaging

box of 10 bottles

bottle capacity

125 mL

bottle filling volume

90 mL

capacity

125 mL

pH

7.3

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

components

lecithin
histidine
cysteine
tween

storage temp.

2-25°C

suitability

selective for (fastidious aerobic bacteria, yeasts, and molds)

General description

Tryptic Soy Broth is a highly nutritious, universal complex medium and hence supports the growth of a wide variety of aerobic and facultative anaerobic microorganisms, including fungi. The neutralizing additives polysorbate (Tween®) 80, sodium thiosulfate, cysteine, histidine, and lecithin reverse the growth inhibitory effect of most disinfectants and antiseptics. Histidine has neutralizing efficiency against formaldehyde and formaldehyde-releasing agents. Sodium thiosulfate inactivates sodium hypochlorite and acidified sodium chlorite. Cysteine is a reducing agent that neutralizes the oxidizing agents such as hydrogen peroxide and iodine. Lecithin, particularly in combination with Polysorbate 80, is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid, and its esters, phenols, phenylethyl alcohols, and Solbrols (PHB esters).

Application

Tryptic Soy Broth with neutralizers is suitable for the isolation and cultivation of aerobic and facultative anaerobic bacteria, yeasts, and molds in the presence of disinfectant residues. The medium is used for the testing of sterile and non-sterile substances, preparations, and products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service