1.46468
Fluid D
ready-to-use, bottle volume 100 mL , filling volume
About This Item
Recommended Products
Agency
EP 2.6.1
JP 4.06
USP 71
71
Quality Level
sterility
sterile; autoclaved
form
liquid
feature
closure type crimp cap with septum
ready-to-use
packaging
bottle of
technique(s)
sterile filtration: suitable
bottle capacity
125 mL
bottle volume
100 mL , filling volume
pH
7.0±0.5
application(s)
cosmetics
food and beverages
pharmaceutical
sterility testing
components
(Sodium Thioglycollate, 1,4-Dithiothreitol)
storage temp.
2-25°C
suitability
nonselective for
General description
Sodium Thioglycollate facilitates the growth ofaerobic and anaerobic microorganisms in the test sample and 1,4-Dithiothreitolensures complete detachment from the sample e.g. prosthetic implant or medicaldevices.
Application
Features and Benefits
- Our ready-to-use mediaprovide the highest level of quality and testing confidence. They areformulated and tested to meet the pharmacopeia requirements.
- Sterility testing media andrinse solutions are manufactured in an ISO 9001, environmentally controlledproduction center. Each lot undergoes a stringent quality control (QC)procedure, including pH, sterility, and growth promotion tests.
- Our manufacturing approachensures the highest level of clarity for our media and rinsing fluids, thusimproving accuracy, and reducing the risk of incorrect interpretation and falseresults.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service