Skip to Content
Merck
All Photos(1)

Key Documents

1.46463

Millipore

Mossel Broth

ready-to-use, bottle filling volume 100 mL

Synonym(s):

Buffered Glucose-Brilliant Green Bile Broth, EE Broth, Enterobacteria Enrichment Broth Mossel

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 100 ML
$112.00

$112.00


Estimated to ship on27 May 2025


Request more information

Select a Size

Change View
10 X 100 ML
$112.00

About This Item

UNSPSC Code:
41106214
NACRES:
NA.74

$112.00


Estimated to ship on27 May 2025


Request more information

Agency

EP 2.6.1
USP

Quality Level

description

For selective enrichment Enterobacteriaceae; Suitable for sterility testing

sterility

sterile; autoclaved

form

liquid

shelf life

8 mo.

feature

closure type blue screw cap with 3 loci
ready-to-use

packaging

single packed of 10 bottles
bottle of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

125 mL

bottle filling volume

100 mL

capacity

125 mL

pH

7.2

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

Enterobacter spp.
nonselective for

General description

The brilliant green and ox bile in the Mossel Broth almost completely inhibit the undesired accompanying bacterial flora. The high buffering capacity of the culture medium prevents the formed acid from killing the culture. Glucose is the main energy source and supports the rapid growth of all Enterobacteriaceae, while enzymatic digest of animal tissue serves as the source for nitrogen, vitamins, and amino acids.

Application

Mossel Broth is suitable for microbiological examination of non-sterile pharmaceutical products, foodstuff and other materials. It is designed for selective enrichment of bile-tolerant, gram-negative bacteria and particularly Enterobacteriaceae.

Features and Benefits

  • Ready-to-use media
  • Bottles with screw caps

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service