Skip to Content
Merck
All Photos(1)

Key Documents

1.46313

Millipore

R2A Agar

Synonym(s):

Reasoner´s 2A agar

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 200 ML
$74.00

$74.00


Estimated to ship on19 May 2025


Request more information

Select a Size

Change View
6 X 200 ML
$74.00

About This Item

UNSPSC Code:
41106204
NACRES:
NA.74

$74.00


Estimated to ship on19 May 2025


Request more information

Agency

APHA
EP
JP

Quality Level

description

For determination of heterotrophic total microbial count

sterility

sterile; autoclaved

form

liquid (ready-to-use)

shelf life

10 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

bottle of
single packed of (200 ml in 250 ml bottle with blue screw cap and 3 loci (6 bottles per box))

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

250 mL

bottle volume

200 mL , Filling volume

pH

7.2

application(s)

cosmetics
food and beverages
pharmaceutical
water monitoring

storage temp.

2-25°C

suitability

nonselective for

General description

R2A Agar (Reasoner´s 2A agar) is designed to promote the growth of micro-organisms that grow in low nutrient environments, such as water. The reduced amount of nutrients in the medium allows the growth of aerobic, heterotrophic micro-organisms, which are adapted to low nutrient content. The low concentration of yeast extract, casein hydrolysate, peptone, and glucose allows the growth of a wide spectrum of micro-organisms, without the fast-growing micro-organisms suppressing the slow-growing species (which would be the case on richly nutritious media). Starch and pyruvate particularly support the growth of injured the micro-organisms.

Application

R2A Agar is used for determining the heterotrophic total microbial count in water for injections in bulk, highly purified water, and purified water in bulk.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Our media are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service