Skip to Content
Merck
All Photos(4)

Key Documents

1.17976

Supelco

Peracetic Acid Test Strips, colorimetric

greener alternative

20-160 mg/L (peracetic acid), MQuant®

Synonym(s):

Peracetic Acid chemical test strips

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$107.00

$107.00


In StockDetails


Request a Bulk OrderRequest more information

Select a Size

Change View
100 TESTS
$107.00

About This Item

UNSPSC Code:
41116116
NACRES:
NB.21

$107.00


In StockDetails


Request a Bulk OrderRequest more information

Product Name

Peracetic Acid Test, colorimetric, 20-160 mg/L (peracetic acid), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

peracetic acid

reaction suitability

reaction type: Redox Reactions

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

20-160 mg/L (peracetic acid)

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric with test strips, 20 - 40 - 80 - 120 - 160 mg/L Peracetic acid, MQuant®

Peracetic acid is a frequently used disinfectant. The MQuant® Peracetic acid test strips are applicable – even in the presence of hydrogen peroxide – to test for the absence of peracetic acid residues after rinsing cycles. Peracetic acid reacts with an aromatic amine to form a blue dye. The concentration of peracetic acid is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the peracetic acid concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service