Skip to Content
Merck
All Photos(1)

Key Documents

1.46237

Millipore

HEIMPLATE Sab-Gluc.(4%)-Agar w. LHT

90 mm Settle Plates, with lecithin, Tween®, and histidine for detection of yeast and molds

Synonym(s):

Sabouraud Dextrose Agar with Lecithin, Tween® and Histidine, SDA with neutralizer

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$54.70

$54.70


Estimated to ship on17 May 2025


Request more information

Select a Size

Change View
20 PKG
$54.70

About This Item

UNSPSC Code:
41106227
NACRES:
NA.74

$54.70


Estimated to ship on17 May 2025


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; aseptically filled

form

plate (ready-to-use)

feature

ready-to-use

technique(s)

microbiological culture: suitable

D

90 mm

pH

5.6 ( in H2O)

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

2-8°C

suitability

molds
yeasts

General description

Sabouraud Glucose Agar + LTH in 90 mm settle plate is designed for the detection of yeasts and molds in the air via active or passive air monitoring as well as fingerprints of personnel in controlled environments in the presence of disinfectant residues. Sabouraud Glucose Medium contains digests of animal tissues (peptones), which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Glucose (4%) is added as the energy and carbon source. The low pH favors fungal growth and inhibits contaminating bacteria from samples. Additionally, the medium is supplemented with neutralizers such as lecithin, polysorbate (Tween®) 80, and histidine to inactivate disinfectant residues.

Application

Sabouraud Glucose Agar + LTH settle plate is designed for the detection of the total count of yeasts and molds in air and from personnel in less critical environments.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service