Key Documents
1.30191
Millipore
HY-LiTE® 2/2A Battery Holder
Replacement battery holder for battery operation of HY-LiTE® 2/2A luminometers
Select a Size
1 UNIT
$266.00
$266.00
Estimated to ship on06 May 2025
Request more information
Select a Size
Change View
1 UNIT
$266.00
About This Item
UNSPSC Code:
41122409
NACRES:
NA.74
$266.00
Estimated to ship on06 May 2025
Request more information
product line
HY-LiTE®
manufacturer/tradename
HY-LiTE®
application(s)
food and beverages
compatibility
for use with HY-LiTE® 2/2A instrument versions.
General description
Replacement or spare battery holder for HY-LiTE® 2/2A instruments. The battery holder is used to operate the luminometer on battery power using 4 sealed alkaline batteries (size Mignon, LR6, AA). (Batteries not included).
Application
For use as a spare part or replacement of the original battery holder included with the luminometer.
Legal Information
HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany