Skip to Content
Merck
All Photos(1)

Documents

1.12400

Sigma-Aldrich

Bismuth

(powder) 99+

Synonym(s):

Bismuth

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
EC Index Number:
231-177-4
NACRES:
NA.22

vapor pressure

1 hPa ( 840 °C)

Quality Level

potency

>2000 mg/kg LD50, oral (Rat)

bp

1564 °C/1013 hPa

mp

271 °C

density

9.78 g/cm3 at 20 °C

bulk density

6000 kg/m3

storage temp.

no temp limit

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Analysis Note

Assay (complexometric): ≥ 99.0 %
Identity (ICP-OES): passes test

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service