Skip to Content
Merck
All Photos(2)

Key Documents

1.19753

Supelco

di-Sodium hydrogen phosphate dihydrate

buffer substance for chromatography LiChropur

Synonym(s):

di-Sodium hydrogen phosphate dihydrate, Sodium monohydrogen phosphate, Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$128.00

$128.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$128.00

About This Item

Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
MDL number:
UNSPSC Code:
12161703
EC Index Number:
231-448-7
NACRES:
NB.21
grade:
reagent grade
form:
solid

$128.00


Check Cart for Availability

Request a Bulk Order

grade

reagent grade

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

technique(s)

HPLC: suitable

pH

9.0-9.4 (20 °C, 50 g/L in H2O)

mp

92.5 °C (Elimination of water of crystallisation)

density

2.1 g/cm3 at 20 °C

bulk density

850‑1000 kg/m3

storage temp.

2-30°C

SMILES string

O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2

InChI key

KDQPSPMLNJTZAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Identity: passes test
pH-value (5 %; water): 9.0 - 9.2
Assay (acidimetric): ≥ 99.0 %
Self-absorption (215 nm; 0.2 mol/l; 1 cm; water): ≤ 0.15
Self-absorption (220 nm; 0.2 mol/l; 1 cm; water): ≤ 0.05
Self-absorption (254 nm; 0.2 mol/l; 1 cm; water): ≤ 0.02

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service