Skip to Content
Merck
All Photos(1)

Key Documents

1.17917

Supelco

Arsenic Test Strips, colorimetric

0.02-3 mg/L (As), MQuant®

Synonym(s):

Arsenic chemical test strips

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$259.00

$259.00


Available to ship on01 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
100 TESTS
$259.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$259.00


Available to ship on01 April 2025Details


Request a Bulk OrderRequest more information

Product Name

Arsenic Test, colorimetric, As 0.02-3 mg/L, MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

arsenic

reaction suitability

reaction type: modified Gutzeit test

As measuring range

0.02-3 mg/L

detection method

colorimetric

storage temp.

15-25°C

General description

Method: Colorimetric with test strips and reagent, 0.02 - 0.05 - 0.1 - 0.2 - 0.5 mg/L As, 0.1 - 0.5 - 1.0 - 1.7 - 3.0 mg/L As, MQuant®

The arsenic test strips are suitable for measuring arsenic in water, pharmaceutical products, prepared biological specimens and liquid foods. Within this MQuant® Arsenic test, test strips are used in combination with additional reagents. Arsenic is liberated from the sample by adding zinc powder, a solid acid, and - for the elimination of interfering sulfide ions - an oxidizing agent. On the reaction zone of the test strip a yellow-brown dye is formed. The concentration of arsenic(III) and arsenic(V) are measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the arsenic concentration and a possible dilution factor prior to using a quantitative method such as our Spectroquant® test.

Application

  • Arsenic Pigments in Libraries – Testing Methods, Contaminated Spaces, and Occupational Safety Measures: In a research project, the Bonn University and State Library and the Cologne University of Applied Sciences analyzed the handling of arsenic in library materials. Arsenic-containing pigments in library materials are a source of danger for institutional staff, conservators, scientists, and users. Over 300 volumes with suspicious green book components were analyzed using both XRF and Raman spectroscopy, with arsenic compounds being detected in around half of them. The XRF device is mobile and effective, it yields reproducible data and can detect a multitude of elements such as arsenic, lead, and mercury. However, it emits radiation and is limited to small spots. Special training for operating the device as well as for interpreting the results is needed. A rapid test based on bromide test strips and the [TM=“MQUANT”] test kit were then evaluated. When comparing the rapid test kits, the [TM=“MQUANT”] test kit appears to be more suitable than the bromide test paper as it includes all necessary accessories and helps minimize the hazard potential for users. The [TM=“MQUANT”] test kit may be suitable for initial in situ examinations, after training and instruction of the archive staff, being fairly accurate, ready-made, easy to handle, and suitable for testing surfaces. (Wetten J et al., 2024)

  • Improved On-Site Characterization of Arsenic in Gypsum from Waste Plasterboards Using Smart Devices.: The research highlights the development of smart device-based arsenic tests for on-site characterization of arsenic in gypsum, enhancing the efficiency and accuracy of environmental monitoring and waste management (Tafu M et al., 2022).

  • MnFe(2)O(4) micromotors enhanced field digestion and solid phase extraction for on-site determination of arsenic in rice and water.: This study employs arsenic tests in conjunction with MnFe(2)O(4) micromotors for on-site arsenic determination in rice and water samples, showcasing advancements in sample preparation and field analysis techniques (Luo Y et al., 2021).

  • Impact of receipt of private well arsenic test results on maternal use of contaminated drinking water in a U.S. population.: The research evaluates the behavioral impact of receiving arsenic test results on maternal use of contaminated drinking water, emphasizing the importance of arsenic testing in public health interventions (He X et al., 2018).

  • Test for arsenic speciation in waters based on a paper-based analytical device with scanometric detection.: This study presents a novel paper-based analytical device for arsenic speciation in water, utilizing scanometric detection to offer a cost-effective and portable solution for environmental monitoring (Pena-Pereira F et al., 2018).

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Would this test work to test taxidermy mounts for arsenic? Could sample hair or feathers be swabbed or left in a solution to then test?

    1 answer
    1. The strips have not been validated for such a method. However, if mounts are swabbed and dispersed into a 10 mL sample, as required by the kit, and the concentration of arsenic in that sample is between 0.02-3 mg/L, then the strips could be useful. Method validation would likely be required to determine feasibility for the end user's mounts and concentrations.

      Helpful?

  2. Hi can we use this test with a hair dye?

    1 answer
    1. This product has not been specifically tested for use with hair dye. However, it is expected to be suitable. Please see the link below to review the user guide:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/272/802/117917ug-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service