Skip to Content
Merck
All Photos(1)

Documents

1.04761

Supelco

Iodine

sublimated for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Iodine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
MDL number:
UNSPSC Code:
12141916
EC Index Number:
231-442-4
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

0.41 hPa ( 25 °C)

product line

EMSURE®

form

solid

potency

14000 mg/kg LD50, oral (Rat)
1425 mg/kg LD50, skin (Rabbit)

pH

5.4 ( in H2O, saturated solution)

kinematic viscosity

0.57 cSt(116 °C) (liquid)

bp

185 °C/1013 hPa

mp

114 °C

solubility

0.3 g/L

density

4.93 g/cm3 at 20 °C

bulk density

2100 kg/m3

storage temp.

2-30°C

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (iodometric): 99.8 - 100.5 %
Identity: passes test
Bromide and Chloride (as Cl): ≤ 0.005 %
Non volatile matter (105 °C): ≤ 0.01 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Protocols

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service