Skip to Content
Merck
All Photos(1)

Key Documents

1.17956

Supelco

Peracetic Acid Test Strips, reflectometric

20.0-100 mg/L (peracetic acid), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Peracetic Acid chemical test strips, Peracetic Acid test strip for instrument reader

Sign Into View Organizational & Contract Pricing

Select a Size

50 TESTS
$84.10

$84.10


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
50 TESTS
$84.10

About This Item

UNSPSC Code:
41116116
NACRES:
NB.21

$84.10


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Product Name

Peracetic Acid Test, reflectometric, 20.0-100 mg/L (peracetic acid), for use with REFLECTOQUANT®

description

quantitative

Quality Level

product line

Reflectoquant®

specific analyte(s)

peracetic acid

reaction suitability

reaction type: Redox Reactions

measuring range

20.0-100 mg/L (peracetic acid)

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 20.0 - 100 mg/l Reflectoquant®
Peracetic acid is a frequently used disinfectant. The Reflectoquant Peracetic acid test strips are suited for the selective determination of the peracetic acid concentration in aqueous solutions and to check for the absence of peracetic acid after rinsing processes, also in cases in which hydrogen peroxide is present. Peracetic acid reacts with a phenol derivative to form a violet dye that is determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to estimate the peracetic acid concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for peracetic acid.

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service