Skip to Content
Merck
All Photos(1)

Key Documents

1.08028

Supelco

Formaldehyde Test Kit, colorimetric

0.1-1.5 mg/L (HCHO), MQuant®

Synonym(s):

Formaldehyde Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$345.00

$345.00


Estimated to ship on11 June 2025FromTruganina


Request a Bulk OrderRequest more information

Select a Size

Change View
100 TESTS
$345.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$345.00


Estimated to ship on11 June 2025FromTruganina


Request a Bulk OrderRequest more information

Product Name

Formaldehyde Test, colorimetric, 0.1-1.5 mg/L (HCHO), for use with MCOLORTEST®

product line

MColortest®

Quality Level

specific analyte(s)

formaldehyde

measuring range

0.1-1.5 mg/L (HCHO)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and sliding comparator 0.1 - 0.25 - 0.4 - 0.6 - 0.8 - 1.0 - 1.5 mg/l HCHO MQuant®
This MQuant® Formaldehyde test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card. The evaluation considers the untreated sample so even turbid and colored water samples can be analyzed without further preparation. Formaldehyde is used as a disinfectant for surfaces, medical equipment and also as a raw material in binding agents, fixing agents or as a preservative.
The test can be used in
  • Disinfectant and rinsing solutions (e. g. laundries)
  • Aqueous solutions
  • Process water
  • Electroplating wastewater
  • Food after appropriate sample pretreatment
This test is not suited for seawater.

Method
Formaldehyde reacts with 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole to form a purple-red tetrazine. The formaldehyde concentration is measured semiquantitatively by allocating the color to a value on a reference scale.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Formaldehyde

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service