Skip to Content
Merck
All Photos(1)

Documents

1.15228

Millipore

N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid

pKa (20 °C)7.15, ≥99% (alkalimetric)

Synonym(s):

N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid, BES

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO5S
CAS Number:
Molecular Weight:
213.25
MDL number:
UNSPSC Code:
12161703
EC Index Number:
233-465-5
NACRES:
NA.05

Quality Level

Assay

≥99% (alkalimetric)

form

solid

pKa (20 °C)

7.15

storage temp.

2-30°C

InChI

1S/C6H15NO5S/c8-4-1-7(2-5-9)3-6-13(10,11)12/h8-9H,1-6H2,(H,10,11,12)

InChI key

AJTVSSFTXWNIRG-UHFFFAOYSA-N

General description

buffer substance BES

Analysis Note

Assay (alkalimetric): ≥ 99 %
Identity (IR-spectrum): passes test
UV-Absorption (250 nm, 50 g/l, 1 cm, water): ≤ 0.05
Chloride (Cl): ≤ 0.005 %
Sulphate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Loss on Drying (105°C): ≤ 0.5 %
Biological buffer, pKa at 20 °C: 7.15

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service