1.14833
Lead Test, photometric
photometric, 0.10-5.00 mg/L (Pb), Spectroquant®
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
Lead Cell Test, photometric, 0.10-5.00 mg/L (Pb), Spectroquant®
product line
Spectroquant®
Quality Level
usage
sufficient for 25 tests
25 tests
specific analyte(s)
lead
measuring range
0.10-5.00 mg/L (Pb)
technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (PAR)
storage temp.
15-25°C
General description
This Spectroquant® Lead Cell Test allows the accurate quantification of the lead(II) ions concentration in aqueous samples. To analyze undissolved or complex-bound lead the sample needs to be decomposed by digestion.
Method applied: In alkaline solution lead(II) ions react with 4-(2’-pyridylazo)resorcinol (PAR) to form a red complex which is determined photometrically.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant®Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 2 - STOT RE 2 Oral
Target Organs
spleen, Thyroid
Supplementary Hazards
Storage Class Code
6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Photometric determination with PAR subsequent to acid mineralisation; DIN decomposition
Photometric determination with PAR subsequent to separation of lead with Sodium diethyldithiocarbaminate
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service