Skip to Content
Merck
All Photos(1)

Key Documents

1.14758

Supelco

Chromate Test, photometric

photometric, 0.010-3.00 mg/L (Cr), 0.02-6.69 mg/L (CrO4), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

250 TESTS
$328.00

$328.00


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
250 TESTS
$328.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$328.00


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

Product Name

Chromate Test, photometric, 0.010-3.00 mg/L (Cr), 0.02-6.69 mg/L (CrO4), Spectroquant®

Quality Level

usage

sufficient for 250 tests

specific analyte(s)

chromate

measuring range

0.010-3.00 mg/L (Cr)
0.02-6.69 mg/L (CrO4)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Diphenylcarbazide)

storage temp.

15-25°C

General description

Method: photometric 0.010 - 3.00 mg/l Cr 0.02 - 6.69 mg/l CrO₄ Spectroquant®

The method is analogous to APHA 3500-Cr D and DIN 38405-52.

This Spectroquant® Chromate Reagent Test allows the accurate quantification the chromium(VI) concentration present in aqueous samples as chromate or dichromate ions. For the detection of complex-bound chromium(III) or total chromium (sum of chromium(VI) and chromium(III)) the sample needs to be decomposed by digestion.

Method applied: In weakly phosphoric solution chromium(VI) ions react with diphenylcarbazide to form chromium(III) and diphenylcarbazone which form a red-violet complex that is determined photometrically.


The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.


All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service