Skip to Content
Merck
All Photos(1)

Key Documents

1.14693

Supelco

PhotoCheck

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$751.00

$751.00


Available to ship on31 March 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
1 SET
$751.00

About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

$751.00


Available to ship on31 March 2025Details


Request a Bulk OrderRequest more information

technique(s)

photometry: suitable

Quality Level

compatibility

for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

storage temp.

15-25°C

General description

Check solutions for photometers (445/446 nm, 520/525 nm, and 690 nm) Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service