Skip to Content
Merck
All Photos(2)

Key Documents

1.10983

Supelco

N,N-Dimethylformamide

for gas chromatography ECD and FID SupraSolv®

Synonym(s):

DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$135.00
2.5 L
$296.00

$135.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$135.00
2.5 L
$296.00

About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-679-5
NACRES:
NA.03
technique(s):
gas chromatography (GC): suitable
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)

$135.00


Available to ship on31 March 2025Details


Request a Bulk Order

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

833 °F

potency

3010 mg/kg LD50, oral (Rat)
1500 mg/kg LD50, skin (Rabbit)

expl. lim.

15.2 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

density

0.944 g/mL (lit.)

storage temp.

2-30°C

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detector) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimum signal-to-noise ratio leading to reliable and reproducible analysis results.

Application

Solvent for many hydrophobic organic compounds.
Used for GC applications such as sensitive detection in residue and environmental analysis.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service