Skip to Content
Merck
All Photos(1)

Key Documents

1.10032

Supelco

Total Hardness Test Strips, colorimetric

individually sealed, water hardness 3-21 °d, water hardness 4-26 °e, water hardness 5-37 °f, MQuant®

Synonym(s):

Water Hardness Chemical Test Strips

Sign Into View Organizational & Contract Pricing

Select a Size

1000 STRIPS
$581.00
25000 PKG
$6,530.00

$581.00


Estimated to ship on17 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1000 STRIPS
$581.00
25000 PKG
$6,530.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$581.00


Estimated to ship on17 May 2025


Request a Bulk OrderRequest more information

Product Name

Total Hardness Test, colorimetric, water hardness 3-21 °d, water hardness 4-26 °e, water hardness 5-37 °f, MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

calcium, magnesium

water hardness measuring range

3-21 °d
4-26 °e
5-37 °f

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with test strips (individually sealed) <3 - >4 - >7 - >14 - >21°d <4 - >5 - >9 - >18 - >26°e <5 - >7 - >12 - >25 - >37°f MQuant®
The total hardness test can be used to determine the water hardness for heating systems, coffee machines, dishwasher and washing machines, allowing to optimize the dosing of water softeners and use of water filters. Water hardness is caused by the presence of alkaline earths, primarily calcium and magnesium. Total hardness is measured as the sum of all calcium and magnesium ions. Total hardness test strips simplify the detection of water hardness by providing a convenient and straightforward testing process on-the-spot. The procedure is simple and convenient, and the clear color changes from green to red provide reliable results within seconds. To use the test strips, simply dip them in the water, wait for the specified time, and then read the results.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service