Skip to Content
Merck
All Photos(1)

Key Documents

1.09446

Supelco

Buffer solution

(citric acid/sodium hydroxide) traceable to SRM from NIST and PTB pH 5.00 (25°C) Certipur®

Synonym(s):

Buffer solution pH 5.00 (25°C) (citric acid/sodium hydroxide)

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$35.00

$35.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$35.00

About This Item

UNSPSC Code:
12161703
NACRES:
NA.25

$35.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

pH

5.0 (25 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The certified buffer solution (citric acid/sodium hydroxide) is gravimetrically prepared using high-quality reagents, in accordance with DIN EN ISO/IEC 17025. Its pH value of 5.00 at 25°C is measured following a 5-point calibration method, using a combined glass electrode. The measurement is made following DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. Its pH value is strongly dependent on the temperature and therefore a constant temperature must be maintained during its use.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The buffer solution is meant to be used as a calibration standard in analytical procedures for pH measuring instruments or pH electrodes and as a control for measuring the pH of solutions.

Features and Benefits

  • Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to secondary reference standard from NIST and primary certified reference standard from PTB
  • Ready-to-use solution
  • Available in a sealed plastic bottle and sustainable Titripac® packaging for a reduced waste generation
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
  • Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
  • Packaging material subjected to comprehensive stability studies as per ISO Guide 35

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service