Skip to Content
Merck
All Photos(1)

Key Documents

1.09434

Supelco

Buffer solution

(citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 3.00 (20°C) Certipur®

Synonym(s):

Buffer solution pH 3.00 (20°C) (citric acid/sodium hydroxide/hydrogen chloride)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$52.40

$52.40


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$52.40

About This Item

UNSPSC Code:
12161704
NACRES:
NA.25

$52.40


Available to ship on02 April 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

vapor pressure

23 hPa ( 20 °C)

product line

Certipur®

pH

3.0 (25 °C in H2O)

bp

101 °C/1013 hPa

mp

-1 °C

density

1.0 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The buffer solution (citric acid/sodium hydroxide/hydrogen chloride) is gravimetrically prepared using high-quality reagents in high-purity water. It is quality controlled and qualified in an ISO/IEC 17025 accredited calibration lab. Its pH value of 3.00 at 20°C is qualified using a combined glass electrode following a 5-point calibration method, in accordance with DIN 19268, against certified primary reference standards as per DIN 19266, IUPAC, NIST, USP, and Ph. Eur.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It is used in analytical procedures for the calibration, qualification, and monitoring of pH measuring instruments and as a control for testing other buffer solutions.

Features and Benefits

  • Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to SRMs from NIST and primary certified reference standards from PTB
  • A ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
  • Long-lasting with a shelf life of 3 years
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service