Skip to Content
Merck
All Photos(1)

Documents

1.01154

Supelco

di-Ammonium hydrogen citrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

di-Ammonium hydrogen citrate, Ammonium citrate dibasic, Diammonium hydrogen citrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O7
CAS Number:
Molecular Weight:
226.18
MDL number:
UNSPSC Code:
12352107
EC Index Number:
221-146-3
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-102.0% (alkalimetric)

form

solid

impurities

≤0.005% Insoluble matter

ign. residue

≤0.01% (as sulfate)

pH

3.3-5.3 (23 g/L in H2O)

density

1.48 g/cm3 at 20 °C

bulk density

400‑600 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
oxalate (C2O42-): passes test (limit about 0.05%)
phosphate (PO43-): ≤0.0005%
sulfur compounds (as SO42-): ≤0.005%

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

InChI

1S/C6H8O7.2H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*1H3

InChI key

YXVFQADLFFNVDS-UHFFFAOYSA-N

Related Categories

Application


  • An EPR and optical absorption study of VO2+ ions in di-ammonium hydrogen citrate [(NH4)2C6H6O7] single crystals.: This research examines the electronic paramagnetic resonance and optical properties of VO2+ ions in di-ammonium hydrogen citrate, which could influence analytical techniques in chemistry and materials science (Karabulut and Tufan, 2008).

Linkage

Replaces: AX1285-1; AX1285; AX1285-3; AX1285-20

Analysis Note

Assay (alkalimetric): 99.0 - 102.0 %
Insoluble matter: ≤ 0.005 %
pH-value (0.1 mol/L): 3.3 - 5.3
Chloride (Cl): ≤ 0.0005 %
Oxalate (C₂O₄): passes test (Limit about 0.05 %)
Phosphate (PO₄): ≤ 0.0005 %
Total sulfur (as SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Residue on ignition (as sulfate): ≤ 0.01 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service