Skip to Content
Merck
All Photos(1)

Key Documents

1.08525

Supelco

Rhodium standard 10 mg/l

(Rhodium(III) nitrate in nitric acid 0.5 mol/l) internal standard for ICP-MS, Certipur®

Synonym(s):

Rhodium internal standard for ICP

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$336.00

$336.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$336.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$336.00


Estimated to ship on22 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

concentration

10 mg/L Rh in 2-3% HNO3

technique(s)

ICP: suitable

pH

1 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Rhodium standard 10 mg/l is a suitable internal standard for elemental analysis by Inductively Coupled Plasma Mass Spectrometry (ICP-MS) technique.

It may serve as an internal standard solution for the ICP-MS analysis:
  • To determine the concentrations of several heavy metals in nail samples[1]
  • To determine uranium in tap and natural water[2]

Analysis Note

10 mg/L Rh in 2-3% nitric acid, prepared with high-purity Rh(NO3)2 and HNO3

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of Cd, Cu, Mn, Ni, Pb and Zn in nail samples by inductively coupled plasma mass spectrometry (ICP-MS) after tetramethylammonium hydroxide solubilization at room temperature: comparison with ETAAS
BL Batista
Talanta, 76, 575-579 (2008)
Determination of uranium in tap water by ICP-MS
ME Himri
Fresenius Journal of Analytical Chemistry, 367, 151-156 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service