Skip to Content
Merck
All Photos(1)

Key Documents

1.08018

Supelco

Color-matching vessel Hydrazine Test Kit, colorimetric

0.1-1.0 mg/L (N₂H₄), MQuant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$250.00

$250.00


Estimated to ship on25 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 UNIT
$250.00

About This Item

UNSPSC Code:
41116116
NACRES:
NB.35

$250.00


Estimated to ship on25 May 2025


Request a Bulk OrderRequest more information

Product Name

Color-matching vessel Hydrazine, 0.1-1.0 mg/L (N2H4), for use with MCOLORTEST®

specific analyte(s)

hydrazine

Quality Level

measuring range

0.1-1.0 mg/L (N2H4)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

We offers you useful accessories. The testing vessel consists of three rectangular chambers that are joined together. These vessels are a necessary accessory for our MQuant® Hydrazine test kit (formally MColortest®) 1.08017 and have to be purchased together with the first test kit.
for 1.08017 0.1 - 0.25 - 0.5 - 1.0 mg/l NH MQuant®

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service