Skip to Content
Merck
All Photos(1)

Key Documents

1.06673

Supelco

Sodium tungstate dihydrate

for analysis EMSURE®

Synonym(s):

Sodium tungstate dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$324.00
1 KG
$811.00
25 KG
$16,200.00

$324.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$324.00
1 KG
$811.00
25 KG
$16,200.00

About This Item

Empirical Formula (Hill Notation):
Na2O4W · 2H2O
CAS Number:
Molecular Weight:
329.85
MDL number:
UNSPSC Code:
12352302
EC Index Number:
236-743-4
NACRES:
NA.21
form:
crystals
solubility:
730 g/L

$324.00


Check Cart for Availability

Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

≥99% (thermogravimetric)

form

crystals

potency

1190 mg/kg LD50, oral (Rat)

pH

8-9 (20 °C, 50 g/L in H2O)

mp

100 °C (Elimination of water of crystallisation)

solubility

730 g/L

density

3.23 g/cm3 at 20 °C

bulk density

1600 kg/m3

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.0005%
Mo: ≤0.005%
N: ≤0.001%
Pb: ≤0.001%
excess alkali (as NaOH): ≤0.10%

storage temp.

15-25°C

SMILES string

[W](=O)(=O)([O-])[O-].[Na+].[Na+].O.O

InChI

1S/2Na.2H2O.4O.W/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;

InChI key

WPZFLQRLSGVIAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Sodium Tungstate Dihydrate (Na2WO4.2H2O): A Mild Oxidizing and Efficient Reagent in Organic Synthesis: This study highlights the use of sodium tungstate dihydrate as a mild and efficient oxidizing agent in organic synthesis. Its applications in various oxidation reactions are discussed, showcasing its versatility and effectiveness in synthesizing complex organic molecules (Jordao et al., 2022).

  • New Findings of Pseudocapacitive Behaviors in Cupric Tungstate Dihydrate: This research uncovers pseudocapacitive properties in cupric tungstate dihydrate, prepared using sodium tungstate dihydrate. The findings contribute to the development of advanced energy storage materials with high capacitance and stability (Gao et al., 2022).

Analysis Note

Assay (thermogravimetric): ≥ 99 %
Identity (x-ray diffraction): passes test
Free alkali (as NaOH): ≤ 0.10 %
Chloride (Cl): ≤ 0.003 %
Sulfate (SO₄): ≤ 0.01 %
Total nitrogen (N): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Mo (Molybdenum): ≤ 0.005 %
Pb (Lead): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service