Skip to Content
Merck
All Photos(1)

Key Documents

1.05694

Supelco

Lithium sulfate monohydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Lithium sulfate monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$277.00

$277.00


Estimated to ship on10 April 2025


Request a Bulk Order

Select a Size

Change View
250 G
$277.00

About This Item

Empirical Formula (Hill Notation):
Li2O4S · xH2O
CAS Number:
Molecular Weight:
109.94 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-820-4
NACRES:
NA.21
Assay:
≥99.0% dry basis (alkalimetric)
grade:
ACS reagent
form:
solid
solubility:
340 g/L

$277.00


Estimated to ship on10 April 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% dry basis (alkalimetric)

form

solid

potency

613 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

loss

13.0-15.0% loss on drying, 150 °C

pH

4.5-6.0 (20 °C, 50 g/L in H2O)

mp

120 °C

solubility

340 g/L

density

2.05 g/cm3 at 20 °C

bulk density

940 kg/m3

anion traces

chloride (Cl-): ≤0.002%
nitrate (NO3-): ≤0.001%

cation traces

Ca: ≤0.005%
Fe: ≤0.0005%
K: ≤0.05%
Na: ≤0.005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Li+].[Li+].[S](=O)(=O)([O-])[O-].O

InChI

1S/2Li.H2O4S.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2

InChI key

RKGLUDFWIKNKMX-UHFFFAOYSA-L

Related Categories

Analysis Note

Assay (alkalimetric, after drying): ≥ 99.0 %
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.002 %
Nitrate (NO₃): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.05 %
Na (Sodium): ≤ 0.005 %
Loss on drying (150 °C): 13.0 - 15.0 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service