Skip to Content
Merck
All Photos(1)

Key Documents

1.06535

Supelco

Sodium nitrate

cryst. EMPLURA®

Synonym(s):

Sodium nitrate, Nitric acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$111.00
50 KG
$3,600.00

$111.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
$111.00
50 KG
$3,600.00

About This Item

Empirical Formula (Hill Notation):
NNaO3
CAS Number:
Molecular Weight:
84.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-554-3
NACRES:
NA.21
form:
solid
solubility:
874 g/L

$111.00


Available to ship on31 March 2025Details


Request a Bulk Order

Quality Level

product line

EMPLURA®

Assay

99.0-100.5% dry basis (acidimetric)

form

solid

potency

1267 mg/kg LD50, oral (Rat)

impurities

≤0.2% Total chlorine

loss

≤2% loss on drying, 105°C

pH

5.5-8.0 (20 °C, 50 g/L in H2O)

mp

308 °C

solubility

874 g/L

density

2.26 g/cm3 at 20 °C

bulk density

1200 kg/m3

anion traces

nitrite (NO2-): ≤0.002%

cation traces

As: ≤0.0003%
Fe: ≤0.001%
Hg: ≤0.0001%
K: ≤0.01%
Pb: ≤0.0002%
Zn: ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].[N+](=O)([O-])[O-]

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Related Categories

Application


  • Nitric oxide pros and cons: The role of L-arginine, a nitric oxide precursor, and idebenone, a coenzyme-Q analogue in ameliorating cerebral hypoxia in rat.: This study discusses the dual roles of nitric oxide and sodium nitrate′s contribution to ameliorating cerebral hypoxia, providing insights into its neurological applications (Abdel Baky NA et al., 2010).

  • Use of experimental design methodology for the development of new magnetic siRNA nanovectors (MSN).: This study details the innovative application of sodium nitrate in the development of magnetic siRNA nanovectors, highlighting its role in enhancing delivery systems (David S et al., 2013).


Analysis Note

Assay (acidimetrisch, after drying): 99.0 - 100.5 %
Identity: passes test
pH-value (5 %; water): 5.5 - 8.3
Total chlorine: ≤ 0.2 %
Nitrite (NO₂): ≤ 0.002 %
As (Arsenic): ≤ 0.0003 %
Fe (Iron): ≤ 0.001 %
Hg (Mercury): ≤ 0.0001 %
K (Potassium): ≤ 0.01 %
Pb (Lead): ≤ 0.0002 %
Zn (Zinc): ≤ 0.001 %
Loss on Drying (105°C): ≤ 2.0 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service