Skip to Content
Merck
All Photos(1)

Key Documents

1.05341

Supelco

Devarda′s alloy

for analysis EMSURE®

Synonym(s):

Devarda′s alloy

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$218.00
1 KG
$529.00

$218.00


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$218.00
1 KG
$529.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352402
NACRES:
NA.21
form:
powder

$218.00


Available to ship on02 April 2025Details


Request a Bulk Order

Quality Level

product line

EMSURE®

form

powder

bp

906 °C/1013 hPa

mp

490-560 °C

density

5.79 g/cm3 at 20 °C

bulk density

2080 kg/m3

cation traces

N: ≤0.001%

storage temp.

2-30°C

SMILES string

[Zn].[Cu].[Al]

InChI

1S/Al.Cu.Zn

InChI key

MUBKMWFYVHYZAI-UHFFFAOYSA-N

Related Categories

General description

Devarda′s alloy is an alloy of aluminium (44% – 46%), copper (49% – 51%), and zinc (4% – 6%). It is used as a reducing agent for the analysis of nitrite and nitrates. Devarda′s alloy EMSURE® grade product) is optimized for regulated analyses and highly demanding lab applications. This reagent is ideal for demanding analytical procedures that need low detection limits and maximum sensitivity.

Features and Benefits

  • Streamlined lab work    
  • Time and cost savings     
  • Superior comparability of results      
  • Certainty during product use      
  • Accuracy regarding impurities      
  • Confidence in analysis and production

Linkage

Replaces: DX0125-2; DX0125

Analysis Note

Total nitrogen (N): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service