Skip to Content
Merck
All Photos(1)

Key Documents

1.05225

Supelco

Cresol red

pH indicator, Reag. Ph Eur

Synonym(s):

Cresol red, o-Cresolsulfonphthalein

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$247.00
25 G
$619.00

$247.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$247.00
25 G
$619.00

About This Item

Empirical Formula (Hill Notation):
C21H18O5S
CAS Number:
Molecular Weight:
382.43
MDL number:
UNSPSC Code:
12171502
EC Index Number:
217-064-2
NACRES:
NA.21

$247.00


Estimated to ship on31 May 2025


Request a Bulk Order

Product Name

Cresol red, indicator Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

solid

loss

≤5% loss on drying, 110°C

visual transition interval

0.5-2.5, pink to brownish-yellow(1st transition range)
6.5-8.5, yellow to purple(2nd transition range)

mp

290 °C (decomposes)

bulk density

600‑620 kg/m3

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)OC(c4c1cccc4)(c3cc(c(cc3)O)C)c2cc(c(cc2)O)C

InChI

1S/C21H18O5S/c1-13-11-15(7-9-18(13)22)21(16-8-10-19(23)14(2)12-16)17-5-3-4-6-20(17)27(24,25)26-21/h3-12,22-23H,1-2H3

InChI key

OBRMNDMBJQTZHV-UHFFFAOYSA-N

Application


  • Description of lipase producing novel yeast species Debaryomyces apis f.a., sp. nov. and a modified pH indicator dye-based method for the screening of lipase producing microorganisms.: This study introduces a novel yeast species, Debaryomyces apis, capable of producing lipase, and a new method utilizing Cresol Red as a pH indicator dye for screening lipase-producing microorganisms (Kumari et al., 2023).

  • Application of colorimetric sensor in monitoring dissolved CO2 in natural waters.: This research discusses the use of a colorimetric sensor, which incorporates Cresol Red, for monitoring dissolved carbon dioxide in natural water sources, providing a method for environmental monitoring and analysis (Lee et al., 2022).

  • Polymer Film Blend of Polyvinyl Alcohol, Trichloroethylene and Cresol Red for Gamma Radiation Dosimetry.: This article presents a polymer film blend incorporating Cresol Red for gamma radiation dosimetry, highlighting its application in monitoring radiation exposure (Doyan et al., 2021).

  • Highly efficient colorimetric CO2 sensors for monitoring CO2 leakage from carbon capture and storage sites.: The study involves the development of highly efficient colorimetric sensors using Cresol Red for detecting CO2 leaks, critical for ensuring safety in carbon capture and storage operations (Ko et al., 2020).



Analysis Note

1st transition range: pH 0.5 - pH 2.5pink - brownish yellow
2nd transition range: pH 6.5 - pH 8.5yellow - purple
Appearance of solution: passes test
Loss on drying (110 °C): ≤ 5
Sensitivity test: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service