Skip to Content
Merck
All Photos(1)

Key Documents

1.02147

Millipore

Contact DC

Envirocheck, Disinfection Control coated with CASO (Tryptic Soy) Agar

Sign Into View Organizational & Contract Pricing

Select a Size

10 UNITS
$58.30

$58.30


Estimated to ship on01 June 2025


Request more information

Select a Size

Change View
10 UNITS
$58.30

About This Item

UNSPSC Code:
41116116
NACRES:
NA.74

$58.30


Estimated to ship on01 June 2025


Request more information

material

plastic cap
plastic slide
plastic tube

Quality Level

product line

Envirocheck

form

solid (ready-to-use)

manufacturer/tradename

Envirocheck

application(s)

food and beverages
surface monitoring
water monitoring

storage temp.

2-30°C

suitability

bacteria (microbial load - before and after disinfection)

General description

(Disinfection Control) Envirocheck
Envirocheck Contact Disinfection Control, a semi-quantitative detection of microorganisms on surfaces and in liquids, are flexible paddles that are coated with Tryptic soy agar to check the effectiveness of disinfection. Slide A is coated with CASO (Tryptic Soy) Agar to determine the total viable (aerobic) count and Slide B is coated with CASO (Tryptic Soy) agar containing neutralizers such as Polysorbat 80, Sodium thiosulphate, Lecithin and Histidine. The use of these neutralizers allows growth of bacteria that were held in bacteriostasis after sanitation/disinfection but still viable

Application

Envirocheck contact slides can be used for both surface and liquid testing.

Analysis Note

Count the number of colonies on both sides of the paddle. CASO Agar: count the number of colonies for obtaining the microbial load before disinfection. CASO Agar with neutralizers: count the number of colonies to obtain the microbial load of surviving microorganisms after disinfection. Growth indicates that disinfection was either not adequate (too low concentration of disinfectant) or the effective time was too short.
Microbial contamination: no growth
pH-value/side one (25 °C): 7.0 - 7.6
pH-value/side two (25 °C): 7.0 - 7.6
Growth/side one
- Escherichia coli ATCC 8739 (WDCM 00012):
good
- Staphylococcus aureus ATCC 6538 (WDCM 00032): good
- Candida albicans ATCC 10231 (WDCM 00054): good
- Bacillus subtilis ATCC 6633 (WDCM 00003): good
- Pseudomonas aeruginosa ATCC 9027 (WDCM 00026): good
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053): good
Growth/side two
- Escherichia coli ATCC 8739 (WDCM 00012):
good
- Staphylococcus aureus ATCC 6538 (WDCM 00032): good
- Candida albicans ATCC 10231 (WDCM 00054): good
- Bacillus subtilis ATCC 6633 (WDCM 00003): good
- Pseudomonas aeruginosa ATCC 9027 (WDCM 00026): good
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053): good
Incubation: 48 hrs.; 35 °C; aerobic; Aspergillus up to 72 hrs.

Legal Information

ENVIROCHECK is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service