Skip to Content
Merck
All Photos(1)

Key Documents

1.01569

Supelco

Chemizorb® Hg

reagent refill pack

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$412.00

$412.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 SET
$412.00

About This Item

UNSPSC Code:
47131905
NACRES:
NA.21

$412.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

form

powder

particle size

<63 μm

storage temp.

15-25°C

General description

Reagents refill pack for Cat. No. 1.12576.0001

Analysis Note

Identity: passes test

Legal Information

CHEMIZORB is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service