Skip to Content
Merck
All Photos(1)

Key Documents

07-839

Sigma-Aldrich

Anti-phospho-IR/IGF1R (Tyr1158) Antibody

Upstate®, from rabbit

Synonym(s):

CD220, Insulin Receptor / CD221, Insulin-like Growth Factor 1 Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$667.00

$667.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$667.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$667.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr1158)

Gene Information

human ... INSR(3643)

General description

diabetes, insulin signaling, glucose

Specificity

Human phospho-Insulin Receptor (IR) Tyrosine 1158 and Insulin-like Growth Factor 1 Receptor (IGF1R) Tyr 1131. Mouse and rat reactivity are expected based on 100% sequence homology.
Predicted to cross-react with mouse and rat based on 100% sequence homology, but have not been tested.

Immunogen

Peptide corresponding to the amino acid region encompassing the human, mouse, and rat phospho-Insulin Receptor (IR) Tyrosine 1158 and Insulin-like Growth Factor 1 Receptor (IGF1R) Tyr 1131.

Application

Detect phospho-IR/IGF1R (Tyr1158) using this Anti-phospho-IR/IGF1R (Tyr1158) Antibody validated for use in IC, IH & WB.
Immunoblot Analysis: A 1:1,000 dilution of this lot detected phospho-IR/IGF1R (Tyr1158) in RIPA lysate of untreated or insulin treated CHO cells transfected with IR.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Target description

155 kDa

Physical form

100 μL of affinity purified rabbit polyclonal IgG in 50% storage buffer (PBS (without Mg2+ and Ca2+), pH 7.3 containing 1.0 mg/mL BSA (IgG, protease free) and 0.05% sodium azide) and 50% glycerol.
Affinity purified

Storage and Stability

Stable for 2 years at -20°C from date of shipment.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerolcontaining solutions to become frozen during storage.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service