Skip to Content
Merck
All Photos(1)

Key Documents

AB5208

Sigma-Aldrich

Anti-Sodium Channel Antibody, Voltage Gated, Brain Type III, pain

Chemicon®, from rabbit

Synonym(s):

Nav1.3, SCN3A

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$813.00

$813.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
$813.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$813.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SCN3A(6328)

Specificity

Recognizes Nav1.3 (type III a subunit of VGSC, Scn3a). Does not cross react with any other sodium channel antigens tested so far.

Immunogen

Purified peptide of alpha subunit (amino acids 511-524) of rat Nav1.3

( type III voltage-gated sodium channel VGSC)

Application

All procedures that are going to receive a full-length protein should be performed at 4°C, and the following protease inhibitor mixture should be used: pepstatin A (1 μg/mL), leupeptin (1 μg/mL), aprotinin (1 μg/mL), Pefabloc SC (0.2 mM), benzamidine (0.1 mg/mL), and calpain inhibitors I and II (8 μg/mL each).

Western blot: 1:200 using ECL on newborn rat brain membranes.

Dilutions should be made using a carrier protein such as BSA (1-3%).

Immunohistochemistry: on fixed frozen sections.

Optimal working dilutions must be determined by the end user.
Detect Sodium Channel using this Anti-Sodium Channel Antibody, Voltage Gated, Brain Type III, pain validated for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA as a stabilizer and 0.05% sodium azide as a preservative. Reconstitute with 50 mL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 0.75 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Gao et al.
BMC neuroscience, 10, 20-20 (2009-03-17)
Taste bud cells transmit information regarding the contents of food from taste receptors embedded in apical microvilli to gustatory nerve fibers innervating basolateral membranes. In particular, taste cells depolarize, activate voltage-gated sodium channels, and fire action potentials in response to
Florian Bolz et al.
Frontiers in neuroanatomy, 11, 28-28 (2017-04-20)
To understand the molecular basis of neuronal excitation in the mammalian olfactory system, we conducted a systematic analysis of the organization of voltage-gated sodium (Nav) channel subunits in the main olfactory epithelium (MOE) and vomeronasal organ (VNO) of adult mice.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service