Skip to Content
Merck
All Photos(4)

Documents

07-729

Sigma-Aldrich

Anti-CTCF Antibody

serum, Upstate®

Synonym(s):

11 zinc finger transcriptional repressor, 11-zinc finger protein, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein), CTCFL paralog, transcriptional repressor CTCF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, canine, rat, primate

packaging

antibody small pack of 25 μL

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CTCF(10664)

General description

CTCF is a ubiquitous 11 zinc finger (ZF) protein with highly versatile functions. A transcriptional repressor, CTCF, binds to promoters of vertebrate c-myc gene. It also binds to the PLK and PIM1 promoters. CTCF may prevent the access of transcriptional activators to enhancers. CTCF also acts as a transcriptional activator of APP. It is involved in different aspects of gene regulation including promoter activation or repression, hormone-responsive gene silencing, methylation-dependent chromatin insulation, and genomic imprinting. CTCF may also act as tumor suppressor. CTCF organizes epigenetically controlled chromatin insulators that regulate imprinted genes in soma.

Specificity

Predicted to cross-react with mouse, rat, dog and primates based on sequence homology.
Recognizes CTCF at a.a. 659-675.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 659-675 (C-TNQPKQNQPTAIIQVED) of human CCCTC-binding factor (CTCF) with a N-terminal cysteine added for conjugation purposes.

Application

Chromatin Immunoprecipitation:
An independent laboratory has confirmed that this antibody immunoprecipitates CTCF-bound chromatin from WEHI B-cells.
Use Anti-CTCF Antibody (Rabbit Polyclonal Antibody) validated in ChIP, WB, ChIP-seq to detect CTCF also known as 11-zinc finger protein, CCCTC-binding factor.

Quality

Routinely evaluated by Western Blot.

Western Blot Analysis:
A 1:1000–1:5000 dilution of this lot detected CTCF in HeLa nuclear extract. A previous lot detected CTCF in K562 nuclear extract (data not shown).

Target description

Approx. 120 kDa

Linkage

Replaces: MABE547

Physical form

Rabbit polyclonal antiserum containing 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C.
Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 µL of extract and boil for 5 minutes to reduce the preparation. Load 20 µg of reduced extract per lane for minigels.

Other Notes

Concentration: The concentration of this serum antibody has not been determined.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of CCCTC binding factor (CTCF) and cohesin in the generation of single-cell diversity of protocadherin-? gene expression.
Monahan, K; Rudnick, ND; Kehayova, PD; Pauli, F; Newberry, KM; Myers, RM; Maniatis, T
Proceedings of the National Academy of Sciences of the USA null
Occupancy of chromatin organizers in the Epstein-Barr virus genome.
Holdorf, MM; Cooper, SB; Yamamoto, KR; Miranda, JJ
Virology null
M D Delgado et al.
FEBS letters, 444(1), 5-10 (1999-02-26)
CTCF is a transcriptional repressor of the c-myc gene. Although CTCF has been characterized in some detail, there is very little information about the regulation of CTCF activity. Therefore we investigated CTCF expression and phosphorylation during induced differentiation of human
Friedreich ataxia patient tissues exhibit increased 5-hydroxymethylcytosine modification and decreased CTCF binding at the FXN locus.
Al-Mahdawi, S; Sandi, C; Mouro Pinto, R; Pook, MA
Testing null
Dysregulated human renin expression in transgenic mice carrying truncated genomic constructs: evidence supporting the presence of insulators at the renin locus.
Zhou, X; Weatherford, ET; Liu, X; Born, E; Keen, HL; Sigmund, CD
American Journal of Physiology: Renal Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service