Skip to Content
Merck
All Photos(1)

Key Documents

07-258

Sigma-Aldrich

Anti-GluR5 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$695.00

$695.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
$695.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$695.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIK1(2897)

Specificity

GluR5 isoforms

Immunogen

peptide corresponding to amino acids 937-949 (CHQRRTQRKETVA) of rat GluR5-2C isoform precursor

Application

Anti-GluR5 Antibody is an antibody against GluR5 for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot on RIPA lysates from a rat brain microsomal preparation

Target description

105-110kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gui-Fang Duan et al.
Nature communications, 9(1), 4879-4879 (2018-11-20)
Kainate-type glutamate receptors play critical roles in excitatory synaptic transmission and synaptic plasticity in the brain. GluK1 and GluK2 possess fundamentally different capabilities in surface trafficking as well as synaptic targeting in hippocampal CA1 neurons. Here we find that the
Scott E Hemby et al.
Journal of neurochemistry, 95(6), 1785-1793 (2005-12-21)
Chronic cocaine and withdrawal induce significant alterations in nucleus accumbens (NAc) glutamatergic function in humans and rodent models of cocaine addiction. Dysregulation of glutamatergic function of the prefrontal cortical-NAc pathway has been proposed as a critical substrate for unmanageable drug
Shuang Qiu et al.
Molecular brain, 11(1), 54-54 (2018-09-23)
Fragile X syndrome is caused by the loss of fragile X mental retardation protein (FMRP). Kainate receptor (KAR) is a subfamily of ionotropic glutamate receptors (iGluR) that acts mainly as a neuromodulator of synaptic transmission and neuronal excitability. However, little

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service