Skip to Content
Merck
All Photos(1)

Documents

07-169-I

Sigma-Aldrich

Anti-iPLA2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

iPLA2, 85/88 kDa calcium-independent phospholipase A2 (EC:3.1.1.4), CaI-PLA2, Group VI phospholipase A2, GVI PLA2, Intracellular membrane-associated calcium-independent phospholipase A2 beta, iPLA2-beta, Patatin-like phospholipase domain-containing prote

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

hamster (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PLA2G6(8398)

General description

Phospholipase A2 (PLA2) enzymes constitute the largest class of phospholipases, members of which catalyze the hydrolysis of the phospholipid sn-2 ester bond, generating a free fatty acid and a lysophospholipid. PLA2 enzymes are divided into 14 groups (I-XIV), including 20 subgroups (e.g. IA, IB, VIA-1, VIA-2, VIB, etc.), based on sequence homology. Historically, PLA2 enzymes have also been classified based on their biological characteristics into three types, secretory (sPLA2), cytosolic Ca2+-dependent (cPLA2), and cytosolic Ca2+-independent (iPLA2), when sequence data were not available. (PMID 12401193 & 12694560). Cat. No. 07-169-I, Anti-iPLA2, is a purified rabbit polyclonal raised against immunogen sequence present in the group VI SH-iPLA2 & LH-iPLA2 (85/88 kDa calcium-independent phospholipase A2) isoforms, but not found in the 53.2 kDa Ankyrin-iPLA2-1 and 46.5 kDa Ankyrin-iPLA2-2 splice variants encoded by the same PLA2G6 (PLPLA9) gene (UniProt O60733). Immunogen sequence is 100% conserved in murine (NP_058611.1) and rodent (AAC53136.1) species.

Immunogen

Epitope: near C-terminal
KLH-conjugated linear peptide corresponding to human iPLA2 near the C-terminal.

Application

Anti-iPLA2 Antibody is an antibody against iPLA2 for use inWestern Blotting and Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected iPLA2 in human tonsil tissue.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected iPLA2 in 10 µg of human testis tissue lysate.

Target description

~70 kDa observed (Ramanadham, S., et al. (2003). Biochemistry. 42(47):13929-13940). This antibody is expected to cross-react to isoforms LH-iPLA2 and SH-iPLA2 only.

Physical form

Affinity
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pancreatic islets and insulinoma cells express a novel isoform of group VIA phospholipase A2 (iPLA2 beta) that participates in glucose-stimulated insulin secretion and is not produced by alternate splicing of the iPLA2 beta transcript.
Ramanadham, S; Song, H; Hsu, FF; Zhang, S; Crankshaw, M; Grant, GA; Newgard, CB; Bao et al.
Biochemistry null
Dayne A Beccano-Kelly et al.
iScience, 26(7), 107044-107044 (2023-07-10)
Parkinson's disease (PD) is characterized by a progressive deterioration of motor and cognitive functions. Although death of dopamine neurons is the hallmark pathology of PD, this is a late-stage disease process preceded by neuronal dysfunction. Here we describe early physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service