Skip to Content
Merck
All Photos(1)

Key Documents

06-1304

Sigma-Aldrich

Anti-SETD8 (hPR-SET7) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone-lysine N-methyltransferase SETD8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$910.00

$910.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$910.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$910.00


Estimated to ship on12 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SETD8(387893)

General description

N-lysine methyltransferase KMT5A (UniProt: Q9NQR1; also known as EC: 2.1.1.43, H4-K20-HMTase KMT5A, Histone-lysine N-methyltransferase KMT5A, Lysine N-methyltransferase 5A, Lysine-specific methylase 5A, PR/SET domain-containing protein 07, PR-Set7, PR/SET07, SET domain-containing protein 8) is encoded by the KMT5A (also known as PRSET7, SET07, SET8, SETD8) gene (Gene ID: 387893) in human. SETD8 (hPR-SET7) is a histone methyltransferase that adds a single methyl group to lysine 20 of histone H4 (H4K20me1). H4K20me1 is enriched during mitosis and represents a specific tag for epigenetic transcriptional repression. SETD8 is not detected during the G1 phase. It is first detected during S through G2 phases, and peaks during mitosis. It mainly functions in euchromatin regions, thereby playing a central role in the silencing of euchromatic genes. It is required for cell proliferation, probably by contributing to the maintenance of proper higher-order structure of DNA during mitosis and is also involved in chromosome condensation and proper cytokinesis. It is also reported to mediate mono-methylation of p53/TP53 at lysine 382, leading to repress p53/TP53-target genes. SETD8 has a SET domain in the amino acids 257-378 and this region contains the active site of enzymatic activity. Sequences upstream and downstream of SET domain are also required for its methyltransferase activity. Two isoforms of SETD8 have been described that are produced by alternative splicing.

Specificity

This rabbit polyclonal antibody detects N-lysine methyltransferase KMT5A (PR-SET7).

Immunogen

GST-tagged recombinant protein corresponding to human SETD8 (hPR-SET7).

Application

Anti-SETD8 (hPR-SET7) Antibody is a Rabbit Polyclonal Antibody for detection of SETD8 (hPR-SET7) also known as Histone-lysine N-methyltransferase SETD8 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blotting in HeLa cell nuclear extract.

Western Blotting Analysis: 0.05 µg/mL of this antibody detected SETD8 (hPR-SET7) in HeLa cell nucleaar extract.

Target description

~43 kDa

Linkage

Replaces: 07-316

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.150 mM NaCl and 0.05% sodium azide.

Storage and Stability

Stable at 2-8°C for 1 year from date of receipt.

Analysis Note

Control
HeLa nuclear extract.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aja Aravamudhan et al.
Physiological reports, 12(7), e15999-e15999 (2024-04-13)
Pulmonary arterial hypertension (PAH) causes pulmonary vascular remodeling, increasing pulmonary vascular resistance (PVR) and leading to right heart failure and death. Matrix stiffening early in the disease promotes remodeling in pulmonary artery smooth muscle cells (PASMCs), contributing to PAH pathogenesis.
Mai Nagasaka et al.
Scientific reports, 13(1), 22877-22877 (2023-12-22)
Transforming growth factor β (TGF-β) is a multifunctional cytokine that induces a diverse set of cellular processes principally through Smad-dependent transcription. Transcriptional responses induced by Smads are tightly regulated by Smad cofactors and histone modifications; however, the underlying mechanisms have
Chieh-Tien Shih et al.
Aging cell, 16(4), 797-813 (2017-05-18)
Cellular senescence is a permanent proliferative arrest triggered by genome instability or aberrant growth stresses, acting as a protective or even tumor-suppressive mechanism. While several key aspects of gene regulation have been known to program this cessation of cell growth
Stine Jørgensen et al.
The Journal of cell biology, 192(1), 43-54 (2011-01-12)
The eukaryotic cell cycle is regulated by multiple ubiquitin-mediated events, such as the timely destruction of cyclins and replication licensing factors. The histone H4 methyltransferase SET8 (Pr-Set7) is required for chromosome compaction in mitosis and for maintenance of genome integrity.
Neysha Tirado-Class et al.
Life science alliance, 7(1) (2023-11-09)
DDB1- and CUL4-associated factors (DCAFs) CDT2 and DCAF14 are substrate receptors for Cullin4-RING E3 ubiquitin ligase (CRL4) complexes. CDT2 is responsible for PCNA-coupled proteolysis of substrates CDT1, p21, and SET8 during S-phase of cell cycle. DCAF14 functions at stalled replication

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service