Skip to Content
Merck
All Photos(1)

Key Documents

07-2143

Sigma-Aldrich

Anti-PLEKHG4B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Pleckstrin homology domain-containing family G member 4B, PH domain-containing family G member 4B

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$509.00

$509.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$509.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$509.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Pleckstrin homology domain-containing family G member 4B (PLEKHG4B) functions as a Rho guanyl-nucleotide exchange factor and is involved in the regulation of Rho protein signal transduction.

Immunogen

GST-tagged recombinant protein corresponding to human PLEKHG4B.

Application

Detect Pleckstrin homology domain-containing family G member 4B using this rabbit polyclonal antibody, Anti-PLEKHG4B Antibody validated for use in western blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in JURKAT cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PLEKHG4B in 10 µg of JURKAT cell lysate.

Target description

~150 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
JURKAT cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service