Skip to Content
Merck
All Photos(1)

Key Documents

05-659

Sigma-Aldrich

Anti-Kaiso Antibody, clone 6F

clone 6F, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$561.00

$561.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
$561.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$561.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6F, monoclonal

species reactivity

mouse, hamster, chicken, canine, rat, human, monkey

manufacturer/tradename

Upstate®

technique(s)

electrophoretic mobility shift assay: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ZBTB33(10009)

Specificity

Kaiso

Immunogen

Recombinant N-terminal GST and His-tagged fusion protein corresponding to residues 103-584 of murine Kaiso

Application

Detect Kaiso with Anti-Kaiso Antibody, clone 6F (Mouse Monoclonal Antibody), that has been shown to work in EMSA, IP, WB & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Immunoblot Analysis: 0.5-2 μg/ml of this antibody detected Kaiso in RIPA lysates from 3T3/A31cells.
Immunohistochemistry: As shown by an independent laboratory, 1μg/ml of this antibody gave positive nuclear immunostaining for Kaiso in NIH 3T3 cells fixed with 100% methanol (Daniel, 2001; Daniel, 1999).
Immunoprecipitation: As shown by an independent laboratory, 2-5μg of this antibody immunoprecipitated Kaiso from RIPA lysates from NIH 3T3 cells (Daniel, 1999; Prokhortchouk, 2001).
Supershift: As shown by an independent laboratory, this antibody will supershift a specific methyl-CpG complex (Prokhortchouk, 2001).

Target description

97kDa

Physical form

0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shun-Dong Dai et al.
Cancer science, 102(1), 95-103 (2010-11-13)
δ-Catenin is the only member of the p120 catenin (p120ctn) subfamily that its primary expression is restricted to the brain. Since δ-catenin is upregulated in human lung cancer, the effects of δ-catenin overexpression in lung cancer still need to be
J M Daniel et al.
Molecular and cellular biology, 19(5), 3614-3623 (1999-04-17)
p120(ctn) is an Armadillo repeat domain protein with structural similarity to the cell adhesion cofactors beta-catenin and plakoglobin. All three proteins interact directly with the cytoplasmic domain of the transmembrane cell adhesion molecule E-cadherin; beta-catenin and plakoglobin bind a carboxy-terminal
Shun-Dong Dai et al.
BMC cancer, 9, 178-178 (2009-06-11)
Kaiso has been identified as a new member of the POZ-zinc finger family of transcription factors that are implicated in development and cancer. Although controversy still exists, Kaiso is supposed to be involved in human cancer. However, there is limited
J M Daniel et al.
Hybridoma, 20(3), 159-166 (2001-07-20)
The POZ-zinc finger protein Kaiso belongs to a rapidly growing superfamily of BTB/POZ zinc finger transcription factors implicated in embryonic development and cancer. Kaiso interacts with the catenin p120(ctn), but the significance of the interaction remains unknown. Although p120(ctn) is
A Prokhortchouk et al.
Genes & development, 15(13), 1613-1618 (2001-07-11)
We describe a novel mammalian DNA binding activity that requires at least two symmetrically methylated CpG dinucleotides in its recognition sequence, preferably within the sequence 5'CGCG. A key component of the activity is Kaiso, a protein with POZ and zinc-finger

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service