Skip to Content
Merck
All Photos(2)

Key Documents

05-184

Sigma-Aldrich

Anti-Src Antibody, clone GD11

clone GD11, Upstate®, from mouse

Synonym(s):

Anti-ASV, Anti-SRC1, Anti-THC6, Anti-c-SRC, Anti-p60-Src

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GD11, monoclonal

species reactivity

avian, mink, mouse, rat, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SRC(6714)

General description

Src is a family of proto-oncogenic tyrosine kinases originally discovered by J. Michael Bishop and Harold E. Varmus. The discovery of Src family proteins has been instrumental to the modern understanding of cancer as a disease where normally healthy cellular signalling has gone awry.
Some tumor-inducing retroviruses (such as Rous sarcoma virus (RSV)), contain a gene called v-src (viral-sarcoma). It was found that the v-src gene in RSV is required for the formation of cancer.
Src tyrosine kinases transmit integrin-dependent signals central to cell movement and proliferation. Hallmarks of v-src induced transformation are rounding of the cell and the formation of actin rich podosomes on the basal surface of the cell. These structures are correlated with increased invasiveness, a process thought to be essential for metastasis.
v-src lacks the C-terminal inhibitory phosphorylation site, and is therefore constitutively active as opposed to normal src (c-src) which is only activated under certain circumstances where it is required (e.g. growth factor signaling). v-src is therefore an instructive example of an oncogene whereas c-src is a proto-oncogene.

Specificity

Recognizes pp60src.

Immunogen

Epitope: amino acids 82-169
Recombinant chicken pp60src; v-src

Application

Anti-Src Antibody, clone GD11 detects level of Src & has been published & validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated on RIPA lysates from non-stimulated A431 carcinoma cells, RIPA lysates from EGF-stimulated A431 cells, mouse 3T3/A31 cells or Cos-1 cells transfected with pUSE Mouse src

Target description

60 kDa

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Control Included: Non-Stim A431 Cell Lysate (12-301). Chicken Embryo Fibroblast (CEF) cells expressing Src protein and plated on fibronectin or Non-stimulated human A431 cell lysate

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Breast Cancer Cells Proliferation Is Regulated by Tyrosine Phosphatase SHP1 through c-jun N-Terminal Kinase and Cooperative Induction of RFX-1 and
Amin S, Kumar A, Nilchi L, Wright K, Kozlowski M
Molecular Cancer Research null
Qiuhua Zhang et al.
Blood, 116(8), 1377-1385 (2010-04-28)
Vascular endothelial growth factor (VEGF) appears to be an important mediator of pathologic retinal angiogenesis. In understanding the mechanisms of pathologic retinal neovascularization, we found that VEGF activates PLD1 in human retinal microvascular endothelial cells, and this event is dependent
Petia P Simeonova et al.
The Journal of biological chemistry, 277(4), 2945-2950 (2001-11-28)
Environmental or occupational exposure to arsenic is associated with a greatly increased risk of skin, urinary bladder, and respiratory tract cancers in arseniasis-endemic areas throughout the world. Arsenic shares many properties of tumor promoters by affecting specific cell signal transduction
Elvira Nieto-Pelegrin et al.
Cell communication and signaling : CCS, 7, 11-11 (2009-05-08)
Cortactin activates the actin-related 2/3 (Arp2/3) complex promoting actin polymerization to remodel cell architecture in multiple processes (e.g. cell migration, membrane trafficking, invadopodia formation etc.). Moreover, it was called the Achilles' heel of the actin cytoskeleton because many pathogens hijack
J Yamauchi et al.
The Journal of biological chemistry, 276(26), 23362-23372 (2001-04-17)
Heterotrimeric G protein G(q) stimulates the activity of p38 mitogen-activated protein kinase (MAPK) in mammalian cells. To investigate the signaling mechanism whereby alpha and betagamma subunits of G(q) activate p38 MAPK, we introduced kinase-deficient mutants of mitogen-activated protein kinase kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service