Skip to Content
Merck
All Photos(3)

Key Documents

05-1106

Sigma-Aldrich

Anti-IGF-IR Antibody, β-subunit, clone 1-2

clone 1-2, from mouse, purified by affinity chromatography

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$632.00

$632.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$632.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$632.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

1-2, monoclonal

purified by

affinity chromatography

species reactivity

mouse, rat

species reactivity (predicted by homology)

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IGF1R(3480)

General description

IGF-I and -II signal through the IGF-I Receptor, which is homologous to the Insulin Receptor. The high-affinity IGF-II Receptor does not play a direct role in signaling, but regulates the concentration of free IGF-II. The IGFs are involved in skeletal growth, and are essential for prevention of apoptosis. Serum levels of free IGFs are kept low by the action of IGF binding proteins (IGFBPs), which sequester the IGFs. Overexpression of IGFBPs may induce apoptosis, presumably by reduction of free IGF; IGFBP levels are also altered in some cancers. The IGF-I Receptor is not as mitogenic as some other growth factor receptors, but its ability to activate the PI3 Kinase pathway, through the Insulin Receptor Substrate (IRS) proteins, is critical for mediating cell survival.

Specificity

Recognizes the β-subunit of type I Insulin-like Growth Factor Receptor (IGF-1R). It shows no cross-reaction with insulin receptor. Its epitope localizes between a.a. 1323-1337 of IGF-1R. Both IGF-1R and insulin receptor are synthesized as a single polypeptide which is glycosylated and proteolytically cleaved to give the α- and β-subunits, which are disulfide-linked in a β-α-α-β configuration in the mature receptor. The α-subunit is completely extracellular, while the β-subunit spans membrane.

Immunogen

A 15-mer peptide corresponding to a.a. 1323-1337 (Tyr-RKNERALPLPQSSTC) from the C-terminal of β-subunit of human IGF-1R.
Epitope: Between a.a. 1323-1337 of IGF-1R

Application

Anti-IGF-IR Antibody, β-subunit, clone 1-2 is an antibody against IGF-IR for use in WB, IH(P).
Immunohistochemistry(paraffin): Representative images from a previous lot.
Optimal Staining of IGF Monoclonal Antibody: Placenta

Western Blot: A previous lot of this antibody was used at 1-2 µg/mL for 2 hours at room temperature.

Optimal working dilutions must be determined by end user.
Research Category
Signaling

Apoptosis & Cancer
Research Sub Category
Insulin/Energy Signaling

Quality

Routinely evaluated by Western Blot on C2C12 lysates.

Western Blot Analysis: 1:500 dilution of this lot detected IGF-1 Receptor beta on 10 μg of C2C12 lysates.

Target description

Recognizes the beta-subunit of type I Insulin-like Growth Factor Receptor (IGF-1R; ~140 kDa)

Linkage

Replaces: MAB1123

Physical form

Affinity purified
Purified mouse monoclonal IgG2b in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.

Storage and Stability

Stable at 2-8°C in undiluted aliquots for up to 1 year from date of receipt.

Analysis Note

Control
C2C12 Cell Lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Sui et al.
Stem cell research & therapy, 11(1), 86-86 (2020-02-28)
Parthenogenetic stem cells (PSCs) are a promising source of regenerated cardiomyocytes; however, their application may be limited without a paternal genome. Insulin-like growth factor-II (IGF-II), a paternally expressed growth hormone, is critical in embryonic differentiation. This study investigated whether forced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service