T-096
Testosterone Calibrator
Level 1, 2 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H28O2
Molecular Weight:
288.42
UNSPSC Code:
12352200
NACRES:
NA.24
Recommended Products
grade
certified reference material
packaging
pkg of 1 mL
manufacturer/tradename
Cerilliant®
IVD
for in vitro diagnostic use
concentration
2 (ng/dL, Stripped Serum)
format
single component solution
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Certified reference material of testosterone in stripped serum, CE marked for in vitro diagnostic (IVD) applications by mass spectrometry. Individual levels of testosterone in stripped serum are offered separately at concentrations ranging from 2 to 2,000 ng/dL. Each individual level is ready to use as a calibrator in LC-MS/MS testosterone methods. Separate options for a blank and 13C- or deuterium labeled internal standards of testosterone or the whole kit including calibrators, blank and Testosterone-2,3,4-13C3 in 10 μg/mL Acetonitrile are also available.
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service