Skip to Content
Merck
All Photos(1)

Key Documents

SCI-001

Supelco

Indeno[1,2,3-cd]pyrene

vial of 1 g, analytical standard, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
1879312
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

packaging

vial of 1 g

manufacturer/tradename

Cerilliant®

application(s)

environmental
forensics and toxicology

format

neat

storage temp.

room temp

SMILES string

c1ccc-2c(c1)-c3ccc4ccc5cccc6cc-2c3c4c56

InChI

1S/C22H12/c1-2-7-17-16(6-1)18-11-10-14-9-8-13-4-3-5-15-12-19(17)22(18)21(14)20(13)15/h1-12H

InChI key

SXQBHARYMNFBPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indeno[1,2,3-cd]pyrene belongs to the class of polycyclic aromatic hydrocarbons (PAHs), also called as polynuclear aromatic hydrocarbons or polyarenes, which are regarded as potential pollutants in the environment. PAHs are typically produced due to natural as well as anthropogenic activities; incomplete combustion of the organic matter being the predominant cause.

Application

Indeno[1,2,3-cd]pyrene may be used as an analytical standard for the quantification of the analyte in sewage sludge, solvent solutions and food extracts, water, and oil samples by various chromatography techniques.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pena-Abaurrea et al.
Journal of chromatography. A, 1256, 222-231 (2012-08-14)
This study evaluates the feasibility of comprehensive two-dimensional gas chromatography coupled to time-of-flight mass spectrometry (GC × GC-ToF MS) for the determination of the 15+1 EU PAHs in sediments. Experimental variables affecting the injection, chromatographic separation and analytical detection of
Walter B Wilson et al.
Analytica chimica acta, 733, 103-109 (2012-06-19)
The analytical performance of BEA - a commercial zeolite - is evaluated for the pre-concentration of fifteen Environmental Protection Agency - polycyclic aromatic hydrocarbons and their subsequent HPLC analysis in tap and lake water samples. The pre-concentration factors obtained with
Elisa D'Alessandro et al.
Cells, 10(11) (2021-11-28)
Coagulation factor (F) Xa induces proinflammatory responses through activation of protease-activated receptors (PARs). However, the effect of FXa on cardiac fibroblasts (CFs) and the contribution of PARs in FXa-induced cellular signalling in CF has not been fully characterised. To answer
Optimization and validation of a new method of analysis for polycyclic aromatic hydrocarbons in sewage sludge by liquid chromatography after microwave assisted extraction
Villar P, et al.
Analytica Chimica Acta, 524(1-2), 295-304 (2004)
Gianfranco Diletti et al.
Journal of chromatography. A, 1062(2), 247-254 (2005-02-01)
A gas chromatographic (GC) method with mass spectrometry detection (MS) for the determination of eight polycyclic aromatic hydrocarbons (PAHs) in olive pomace oil has been developed. The oil was diluted with n-pentane and extracted by liquid-liquid partition with dimethyl sulphoxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service