Skip to Content
Merck
All Photos(1)

Key Documents

ERD-051

Supelco

Dibenzo[a,l]pyrene

vial of 25 mg, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H14
CAS Number:
Molecular Weight:
302.37
Beilstein:
2054068
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

packaging

vial of 25 mg

manufacturer/tradename

Cerilliant®

application(s)

environmental
forensics and toxicology

format

neat

storage temp.

room temp

SMILES string

c1ccc2c(c1)cc3ccc4cccc5c6ccccc6c2c3c45

InChI

1S/C24H14/c1-2-8-18-16(6-1)14-17-13-12-15-7-5-11-20-19-9-3-4-10-21(19)24(18)23(17)22(15)20/h1-14H

InChI key

JNTHRSHGARDABO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibenzo[a,l]pyrene is a polycyclic aromatic hydrocarbon (PAH) found in cigarette smoke condensate and coal combustion. It is an environmental pollutant and a potent carcinogen known to induce tumors in rat mammary gland, mouse skin, lungs and liver.

Application

Dibenzo[a,l]pyrene may be used as an analytical standard for the determination of the analyte in crude extracts of environmental samples, water samples and Chinese coal fly ash CRM by chromatography and spectrometry techniques.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of benzo [a] pyrene and dibenzopyrenes in a Chinese coal fly ash certified reference material
Masala S, et al.
The Science of the Total Environment, 432, 97-102 (2012)
Direct determination of dibenzo [a, l] pyrene in crude extracts of environmental samples by laser-excited Shpol'skii spectroscopy
Kozin IS, et al.
Analytical Chemistry, 67(9), 1623-1626 (1995)
Direct Determination of Dibenzo [a, l] pyrene and Its Four Dibenzopyrene Isomers in Water Samples by Solid- Liquid Extraction and Laser-Excited Time-Resolved Shpol'skii Spectrometry
Yu S and Campiglia AD
Analytical Chemistry, 77(5), 1440-1447 (2005)
The potent carcinogen dibenzo [a, l] pyrene is metabolically activated to fjord-region 11, 12-diol 13, 14-epoxides in human mammary carcinoma MCF-7 cell cultures
Ralston SL, et al.
Cancer Research, 54(4), 887-890 (1994)
George S Bailey et al.
Chemical research in toxicology, 22(7), 1264-1276 (2009-05-20)
Assessment of human cancer risk from animal carcinogen studies is severely limited by inadequate experimental data at environmentally relevant exposures and by procedures requiring modeled extrapolations many orders of magnitude below observable data. We used rainbow trout, an animal model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service