Skip to Content
Merck
All Photos(1)

Key Documents

A-196

Supelco

8-Aminoclonazolam-D4 solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

6-(2-chlorophenyl-3,4,5,6-d4)-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepin-8-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H10D4ClN5
Molecular Weight:
327.80
UNSPSC Code:
41116107
NACRES:
NA.24
Product A-196 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

-65 to -96°C

General description

Snap-N-Spike®/Snap-N-Shoot®
This stable-labeled internal standard is suitable for quantification of 8-aminoclonazolam by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications. 8-Aminoclonazolam is a benzodiazepine metabolite of clonazolam.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service