Key Documents
BR36401
BRAND® BLAUBRAND® volumetric flask trapezoidal
capacity 1 mL, polypropylene stopper
Synonym(s):
BLAUBRAND®, Volumetric flasks
Select a Size
2 EA
$67.00
$67.00
Available to ship on28 April 2025Details
Select a Size
Change View
2 EA
$67.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$67.00
Available to ship on28 April 2025Details
material
polypropylene stopper
transparent borosilicate glass 3.3 flask
Joint
joint ST/NS neck7/16
feature
accuracy class A
packaging
pack of 2 ea
manufacturer/tradename
BRAND 36401
capacity
1 mL
General description
Borosilicate 3.3. To DIN EN ISO 1042. Calibrated to contain (TC, In).
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)