Skip to Content
Merck
All Photos(1)

Key Documents

LM2511

Avanti

Glucosyl (β) C12 Ceramide

Avanti Research - A Croda Brand LM2511, ethanol solution

Synonym(s):

N-(dodecanoyl)-1-β-glucosyl-sphing-4-ene

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$328.00

$328.00


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$328.00

About This Item

Empirical Formula (Hill Notation):
C36H69NO8
CAS Number:
Molecular Weight:
643.93
UNSPSC Code:
12352211
NACRES:
NA.25

$328.00


Available to ship on01 April 2025Details


Request a Bulk Order

form

ethanol solution

packaging

pkg of 1 × 1 mL (LM2511-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM2511

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O

InChI

1S/C36H69NO8/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-30(39)29(28-44-36-35(43)34(42)33(41)31(27-38)45-36)37-32(40)26-24-22-20-17-12-10-8-6-4-2/h23,25,29-31,33-36,38-39,41-43H,3-22,24,26-28H2,1-2H3,(H,37,40)/b25-23+/t29-,30+,31?,33+,34?,35?,36+/m0/s1

InChI key

IYCYEZLMOLRFAN-QGJHTTTBSA-N

Application

Lactosyl (β) C12 Ceramide is suitable as an internal standard:
  • to determine if the isopropanol is a choice procedure for large-scale lipid profiling using UHPLC-MS[1]
  • to assess the effects of diabetes and the treatment with dihydroprogesterone (DHP) in rat cerebral cortex[2]
  • in the lipidomic analysis of extracellular vesicles (EVs) released by human fibroblasts undergoing Oncogene-Induced Senescence (OIS)[3]

Packaging

2 mL Amber Glass Sealed Ampule (LM2511-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isopropanol extraction for cerebrospinal fluid lipidomic profiling analysis
Iriondo A, et al.
Talanta, 195, 619-627 (2019)
Diabetes alters myelin lipid profile in rat cerebral cortex: Protective effects of dihydroprogesterone
Cermenati G, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 168, 60-70 (2017)
Extracellular vesicles released by fibroblasts undergoing H-Ras induced senescence show changes in lipid profile
Buratta S, et al.
PLoS ONE, 12(11), e0188840-e0188840 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service