Skip to Content
Merck
All Photos(1)

Key Documents

LM1903

Avanti

17:0-20:4 PI(3,4)P2

Avanti Research - A Croda Brand LM1903, powder

Synonym(s):

1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′-bisphosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$640.00

$640.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$640.00

About This Item

Empirical Formula (Hill Notation):
C46H92N3O19P3
CAS Number:
Molecular Weight:
1084.15
UNSPSC Code:
41116107
NACRES:
NA.25

$640.00


Check Cart for Availability

Request a Bulk Order

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand LM1903

packaging

1 ea of 1 × (with stopper and crimp cap (LM1903-1EA))

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](OP([O-])(O)=O)[C@@H](OP(O)([O-])=O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+].[NH4+]

InChI

1S/C46H83O19P3.3H3N/c1-3-5-7-9-11-13-15-17-19-20-21-23-25-27-29-31-33-35-40(48)62-38(36-60-39(47)34-32-30-28-26-24-22-18-16-14-12-10-8-6-4-2)37-61-68(58,59)65-44-41(49)42(50)45(63-66(52,53)54)46(43(44)51)64-67(55,56)57;;;/h11,13,17,19,21,23,27,29,38,41-46

InChI key

OKMXNUJCHZSHOL-HTJGCCCZSA-N

Biochem/physiol Actions

Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) is a signaling molecule, which might promote actin aggregation and neuritogenesis.[1] PI(3,4)P2 is associated with proteins and enzymes involved in signal transduction and the cytoskeleton. It is essential for the enzyme action of phospholipase D and ADP-ribosylation factor (ARF).[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (LM1903-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjeev Sharma et al.
Cell reports, 27(7), 1979-1990 (2019-05-16)
Phosphatidylinositol 3,4,5-trisphosphate (PIP3) generation at the plasma membrane is a key event during activation of receptor tyrosine kinases such as the insulin receptor required for normal growth and metabolism. We report that in Drosophila, phosphatidylinositol 5 phosphate 4-kinase (PIP4K) is
Shu-Xin Zhang et al.
Cell research, 27(2), 253-273 (2017-01-21)
Neurite initiation is critical for neuronal morphogenesis and early neural circuit development. Recent studies showed that local actin aggregation underneath the cell membrane determined the site of neurite initiation. An immediately arising question is what signaling mechanism initiated actin aggregation.
Maria Saliakoura et al.
Oncogene, 39(14), 2948-2960 (2020-02-09)
Enhanced prostaglandin production promotes the development and progression of cancer. Prostaglandins are generated from arachidonic acid (AA) by the action of cyclooxygenase (COX) isoenzymes. However, how cancer cells are able to maintain an elevated supply of AA for prostaglandin production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service